Learn More
Apexbio Technology LLC Azithromycin Dihydrate 117772-70-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092617
Encompass_Preferred
Azithromycin Dihydrate (CAS 117772-70-0) is a macrolide antibiotic targeting the bacterial 50S ribosomal subunit It is designed to inhibit bacterial protein synthesis thereby disrupting bacterial growth and replication Azithromycin Dihydrate exerts its biological activity primarily through reversible binding to the 50S ribosomal subunit inhibiting protein synthesis In in vitro assays Azithromycin Dihydrate demonstrates inhibitory activity against susceptible bacterial strains with reported IC50 values typically in the micromolar range Based on these pharmacological properties Azithromycin Dihydrate holds research potential in investigating mechanisms of antibiotic resistance bacterial susceptibility and developing therapeutic strategies against bacterial infections
Catalog No. 50-000-92617 Supplier Apexbio Technology LLC Supplier No. B16695.1
Azithromycin Dihydrate (CAS 117772-70-0) is a macrolide antibiotic targeting the bacterial 50S ribosomal subunit It is designed to inhibit bacterial protein synthesis thereby disrupting bacterial growth and replication Azithromycin Dihydrate exerts its biological activity primarily through reversible binding to the 50S ribosomal subunit inhibiting protein synthesis In in vitro assays Azithromycin Dihydrate demonstrates inhibitory activity against susceptible bacterial strains with reported IC50 values typically in the micromolar range Based on these pharmacological properties Azithromycin Dihydrate holds research potential in investigating mechanisms of antibiotic resistance bacterial susceptibility and developing therapeutic strategies against bacterial infections