Learn More
Selleck Chemical LLC Azithromycin Dihydrate S4147-25mg
SDP
Catalog No. 503108901
Encompass_Preferred
Azithromycin Dihydrate(CP-62993 dihydrate) is an acid stable orally administered macrolide antimicrobial drug structurally related to erythromycin
Catalog No. 50-310-8901 Supplier Selleck Chemical LLC Supplier No. S414725MG
Azithromycin Dihydrate(CP-62993 dihydrate) is an acid stable orally administered macrolide antimicrobial drug structurally related to erythromycin