Learn More
Selleck Chemical LLC Azithromycin Dihydrate S4147-1g
SDP
Catalog No. 503109100
Encompass_Preferred
Azithromycin Dihydrate(CP-62993 dihydrate) is an acid stable orally administered macrolide antimicrobial drug structurally related to erythromycin
Catalog No. 50-310-9100 Supplier Selleck Chemical LLC Supplier No. S41471G
Azithromycin Dihydrate(CP-62993 dihydrate) is an acid stable orally administered macrolide antimicrobial drug structurally related to erythromycin