Learn More
Apexbio Technology LLC Azithromycin Dihydrate 117772-70-0 25mg
SDP
Catalog No. 502240463
Encompass_Preferred
Azithromycin Dihydrate (CAS 117772-70-0) is a macrolide antibiotic targeting the bacterial 50S ribosomal subunit It is designed to inhibit bacterial protein synthesis thereby disrupting bacterial growth and replication Azithromycin Dihydrate exerts its biological activity primarily through reversible binding to the 50S ribosomal subunit inhibiting protein synthesis In in vitro assays Azithromycin Dihydrate demonstrates inhibitory activity against susceptible bacterial strains with reported IC50 values typically in the micromolar range Based on these pharmacological properties Azithromycin Dihydrate holds research potential in investigating mechanisms of antibiotic resistance bacterial susceptibility and developing therapeutic strategies against bacterial infections
This item is not returnable. View return policy
Catalog No. 50-224-0463 Supplier Apexbio Technology LLC Supplier No. B166925
Azithromycin Dihydrate (CAS 117772-70-0) is a macrolide antibiotic targeting the bacterial 50S ribosomal subunit It is designed to inhibit bacterial protein synthesis thereby disrupting bacterial growth and replication Azithromycin Dihydrate exerts its biological activity primarily through reversible binding to the 50S ribosomal subunit inhibiting protein synthesis In in vitro assays Azithromycin Dihydrate demonstrates inhibitory activity against susceptible bacterial strains with reported IC50 values typically in the micromolar range Based on these pharmacological properties Azithromycin Dihydrate holds research potential in investigating mechanisms of antibiotic resistance bacterial susceptibility and developing therapeutic strategies against bacterial infections