Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azithromycin Dihydrate, G-Biosciences
SCI_ED

Catalog No. S139744
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139744 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139744 Supplier G-Biosciences Supplier No. RC890
Add to Cart
Add to Cart

Azithromycin is a broad-spectrum macrolide antibiotic similar in structure to erythromycin. It is effective against various gram-negative and gram-positive bacteria.

It has also displayed efficacy against certain Mycoplasma species. Azithromycin also exhibits anti-inflammatory and immunosuppressive properties.

Chemical Identifiers

CAS 117772-70-0

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 25 mg
Percent Purity >98%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.