Learn More
Medchemexpress LLC Azithromycin hydrate | 117772-70-0 | 785.02 | 5 MG
SDP
Catalog No. 5000386383
Encompass_Preferred
Azithromycin hydrate is a macrolide antibiotic useful for the treatment of a number of bacterial infections.
  • Useful for bacterial infections.
  • Augments rhinovirus-induced IFNβ expression.
  • Specifically reduces MMP-9 mRNA and protein levels.
  • No effect on bronchoalveolar lavage inflammatory parameters in mouse model of asthma exacerbation.
  • Augments expression of interferon-stimulated genes and pattern recognition receptor MDA5.
Catalog No. 50-003-86383 Supplier Medchemexpress LLC Supplier No. HY17506AR5MG
Azithromycin hydrate is a macrolide antibiotic useful for the treatment of a number of bacterial infections.
  • Useful for bacterial infections.
  • Augments rhinovirus-induced IFNβ expression.
  • Specifically reduces MMP-9 mRNA and protein levels.
  • No effect on bronchoalveolar lavage inflammatory parameters in mouse model of asthma exacerbation.
  • Augments expression of interferon-stimulated genes and pattern recognition receptor MDA5.