Learn More
Medchemexpress LLC Azithromycin hydrate | 117772-70-0 | 98.0% | 785.02 | 1 ML
SDP
Catalog No. 5000386920
Encompass_Preferred
Azithromycin hydrate is a macrolide antibiotic effective for treating various bacterial infections. It has been shown to augment rhinovirus-induced IFNβ expression and reduce viral replication in primary bronchial epithelial cells. It also specifically reduces MMP-9 mRNA and protein levels. This product is for research use only.
  • Useful for the treatment of a number of bacterial infections.
  • Augments rhinovirus-induced IFNβ expression.
  • Reduces viral replication.
  • Reduces MMP-9 mRNA and protein levels.
  • Induces expression of interferon-stimulated genes.
Catalog No. 50-003-86920 Supplier Medchemexpress LLC Supplier No. HY17506A10MM/1ML
Azithromycin hydrate is a macrolide antibiotic effective for treating various bacterial infections. It has been shown to augment rhinovirus-induced IFNβ expression and reduce viral replication in primary bronchial epithelial cells. It also specifically reduces MMP-9 mRNA and protein levels. This product is for research use only.
  • Useful for the treatment of a number of bacterial infections.
  • Augments rhinovirus-induced IFNβ expression.
  • Reduces viral replication.
  • Reduces MMP-9 mRNA and protein levels.
  • Induces expression of interferon-stimulated genes.