Learn More
Medchemexpress LLC Azithromycin (hydrate) | 117772-70-0 | 98.0% | 785.02 | 100 MG
SDP
Catalog No. 5000386789
Encompass_Preferred
Azithromycin hydrate is a macrolide antibiotic effective for treating a number of bacterial infections. It is intended for laboratory research use only.
  • Effective macrolide antibiotic
  • Useful for treatment of bacterial infections
  • Enhances rhinovirus-induced IFNβ expression
  • Represses viral replication
  • Lowers MMP-9 mRNA and protein levels in THP-1 cells
  • For laboratory research use only
Catalog No. 50-003-86789 Supplier Medchemexpress LLC Supplier No. HY17506A100MG
Azithromycin hydrate is a macrolide antibiotic effective for treating a number of bacterial infections. It is intended for laboratory research use only.
  • Effective macrolide antibiotic
  • Useful for treatment of bacterial infections
  • Enhances rhinovirus-induced IFNβ expression
  • Represses viral replication
  • Lowers MMP-9 mRNA and protein levels in THP-1 cells
  • For laboratory research use only