Learn More
Medchemexpress LLC Azithromycin (hydrate) | 117772-70-0 | 98.0% | 785.02 | 25 MG
SDP
Catalog No. 5000384461
Encompass_Preferred
Azithromycin hydrate is a macrolide antibiotic effective against various bacterial infections. This compound has also demonstrated antiviral activity against HCoV-229E and HCoV-OC43 in vitro, and exhibits immunomodulatory effects by enhancing rhinovirus-induced IFNβ expression and lowering MMP-9 levels. It is also explored for its potential in autophagy pathways and as an antiparasitic agent. Intended for research use only, this product is not for medical or patient use.
  • Functions as a macrolide antibiotic
  • Shows antiviral activity against specific coronaviruses
  • Enhances antiviral immune responses in bronchial epithelial cells
  • Reduces inflammatory markers like MMP-9 levels
  • Has potential applications in autophagy and as an antiparasitic
  • Available for various research applications and compound screening
  • Used in numerous clinical trials for diverse conditions
Catalog No. 50-003-84461 Supplier Medchemexpress LLC Supplier No. 25MGHY17506AR
Azithromycin hydrate is a macrolide antibiotic effective against various bacterial infections. This compound has also demonstrated antiviral activity against HCoV-229E and HCoV-OC43 in vitro, and exhibits immunomodulatory effects by enhancing rhinovirus-induced IFNβ expression and lowering MMP-9 levels. It is also explored for its potential in autophagy pathways and as an antiparasitic agent. Intended for research use only, this product is not for medical or patient use.
  • Functions as a macrolide antibiotic
  • Shows antiviral activity against specific coronaviruses
  • Enhances antiviral immune responses in bronchial epithelial cells
  • Reduces inflammatory markers like MMP-9 levels
  • Has potential applications in autophagy and as an antiparasitic
  • Available for various research applications and compound screening
  • Used in numerous clinical trials for diverse conditions