Learn More
Medchemexpress LLC Azithromycin hydrate | 117772-70-0 | 98.0% | 785.02 | 50 MG
SDP
Catalog No. 5000386382
Encompass_Preferred
Azithromycin hydrate is a macrolide antibiotic useful for the treatment of various bacterial infections. It is intended for research use only.
  • Macrolide antibiotic.
  • Useful for treating various bacterial infections.
  • Enhances rhinovirus-induced IFNβ expression in primary bronchial epithelial cells.
  • Reduces MMP-9 mRNA and protein levels.
  • Soluble in DMSO.
  • Augments expression of interferon-stimulated genes and MDA5.
  • Intended for research use only.
Catalog No. 50-003-86382 Supplier Medchemexpress LLC Supplier No. HY17506AR50MG
Azithromycin hydrate is a macrolide antibiotic useful for the treatment of various bacterial infections. It is intended for research use only.
  • Macrolide antibiotic.
  • Useful for treating various bacterial infections.
  • Enhances rhinovirus-induced IFNβ expression in primary bronchial epithelial cells.
  • Reduces MMP-9 mRNA and protein levels.
  • Soluble in DMSO.
  • Augments expression of interferon-stimulated genes and MDA5.
  • Intended for research use only.