Learn More
Medchemexpress LLC Azithromycin (hydrate) | 117772-70-0 | MFCD01862248 | 99.9% | 785.02 g/mol | C38H76N2O14 | 10 MG
SDP
Catalog No. 5000299197
Encompass_Preferred
Azithromycin hydrate is the hydrate form of the macrolide antibiotic azithromycin supplied as an analytical standard for laboratory and research use. It is a solid hydrate with CAS 117772-70-0, molecular formula C38H76N2O14, and molecular weight 785.02 g/mol. Intended for analytical assays, method development, and quality control; not for human therapeutic use.
  • High purity (99.85% by HPLC).
  • Analytical assay reported for batch qualification.
  • Solid hydrate form suitable for storage and handling.
  • Available in small standard package sizes for analytical workflows.
  • Suitable for chromatography and mass spectrometry reference standards.
  • SDS and COA available to support handling and traceability.
Catalog No. 50-002-99197 Supplier Medchemexpress LLC Supplier No. HY17506AR10MG
Azithromycin hydrate is the hydrate form of the macrolide antibiotic azithromycin supplied as an analytical standard for laboratory and research use. It is a solid hydrate with CAS 117772-70-0, molecular formula C38H76N2O14, and molecular weight 785.02 g/mol. Intended for analytical assays, method development, and quality control; not for human therapeutic use.
  • High purity (99.85% by HPLC).
  • Analytical assay reported for batch qualification.
  • Solid hydrate form suitable for storage and handling.
  • Available in small standard package sizes for analytical workflows.
  • Suitable for chromatography and mass spectrometry reference standards.
  • SDS and COA available to support handling and traceability.