Learn More
Cayman Chemical Azlocillin (sodium salt), Purity: ≥95%, CAS: 37091-65-9, 500 mg
SDP
Catalog No. 502965732
Encompass_Preferred
A semisynthetic with broad spectrum antibacterial properties; used as part of antibiotic cocktails known as PANTA (with polymyxin B, amphotericin B, nalidixic acid, and trimethoprim) and PANTAV (PANTA with vancomycin)
Catalog No. 50-296-5732 Supplier Cayman Chemical Supplier No. 18424500 MG
A semisynthetic with broad spectrum antibacterial properties; used as part of antibiotic cocktails known as PANTA (with polymyxin B, amphotericin B, nalidixic acid, and trimethoprim) and PANTAV (PANTA with vancomycin)