Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azlocillin sodium, 90%, Thermo Scientific™

Catalog No. AAJ6689509
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6689509 10 g
AAJ6689503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6689509 Supplier Thermo Scientific Chemicals Supplier No. J6689509

missing translation for 'validMainframeMsg'

An extended spectrum --lactam antibiotic derived from ampicillin

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37091-65-9
Molecular Formula C20H22N5NaO6S
Molecular Weight (g/mol) 483.475
MDL Number MFCD07793330
InChI Key UVOCNBWUHNCKJM-XPOQXDFJSA-M
PubChem CID 134129665
IUPAC Name sodium;(2R,5S,6R)-3,3-dimethyl-7-oxo-6-[[(2R)-2-[(2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)NC(=O)N4CCNC4=O)C(=O)[O-])C.[Na+]

Specifications

Type Azlocillin Sodium
Physical Form Powder
Packaging Plastic Bottle
Quantity 10 g
Flash Point >110°C (230°F)
Synonym D-^a-([Imidazolidin-2-on-1-yl]carbonylamino)benzylpenicillin sodium salt
Solubility Information Freely soluble in water
Formula Weight 483.47
Percent Purity 95%
Chemical Name or Material Azlocillin sodium
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 253-347-7
RTECSNumber XH9250000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.