Learn More
Apexbio Technology LLC Azlocillin sodium salt 37091-65-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B18935.1
Encompass_Preferred
Azlocillin sodium salt (CAS 37091-65-9) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis thereby impeding cross-linking of peptidoglycan layers and inducing bacterial lysis Azlocillin sodium salt exerts its biological activity primarily through inhibition of PBPs disrupting cell wall integrity in bacteria In microbiological assays azlocillin demonstrates broad-spectrum antibacterial activity against both Gram-positive and Gram-negative bacteria reported IC50 values vary by microbial species and experimental conditions typically determined in minimum inhibitory concentration (MIC) assays Based on these pharmacological properties azlocillin sodium salt holds research potential in microbiological and biomedical investigations including studies of bacterial susceptibility resistance mechanisms and combinational antimicrobial therapies
Catalog No. 50-000-89975