Learn More
Medchemexpress LLC Azlocillin (sodium salt) | 37091-65-9 | 99.1% | 10 MM 1 ML
SDP
Catalog No. 5000405557
Encompass_Preferred
Azlocillin sodium salt is a semisynthetic penicillin and a broad-spectrum β-lactam antibiotic. It exhibits antipseudomonal activity and is potent against the malarial parasite Plasmodium falciparum.
  • Inhibits over 75% of Pseudomonas aeruginosa isolates
  • Active against indole-negative and -positive Proteus species
  • Effective against gram-positive cocci (except penicillin G-resistant Staphylococcus aureus)
  • Increases survival of neutropenic mice infected with Escherichia coli or Klebsiella pneumoniae
Catalog No. 50-004-05557 Supplier Medchemexpress LLC Supplier No. HYB0529A10MM/1ML
Azlocillin sodium salt is a semisynthetic penicillin and a broad-spectrum β-lactam antibiotic. It exhibits antipseudomonal activity and is potent against the malarial parasite Plasmodium falciparum.
  • Inhibits over 75% of Pseudomonas aeruginosa isolates
  • Active against indole-negative and -positive Proteus species
  • Effective against gram-positive cocci (except penicillin G-resistant Staphylococcus aureus)
  • Increases survival of neutropenic mice infected with Escherichia coli or Klebsiella pneumoniae