Learn More
Cole-Parmer Azlon 522085-0500 polypropylene "square ratio" beaker, 500 mL
Catalog No. NC0434847 Shop All Cole-Parmer Products
Encompass_Preferred
Polypropylene (PP) "square ratio" beakers feature a vertical lip that serves two purposes. First, the lip contains spills that may occur with rounded lips. Second, this vertical lip allows the beakers to be stacked together in sets for easy storage. Maximum temperature for PP is 135°C.
Catalog No. NC0434847 Supplier Cole-Parmer Supplier No. 6620107
Polypropylene (PP) "square ratio" beakers feature a vertical lip that serves two purposes. First, the lip contains spills that may occur with rounded lips. Second, this vertical lip allows the beakers to be stacked together in sets for easy storage. Maximum temperature for PP is 135°C.