Learn More
Cole-Parmer Azlon 537835-1000 Squat-Form PP Graduated Cylinders, 1000 mL, 1/Pk
Catalog No. NC0852346 Shop All Cole-Parmer Products
Encompass_Preferred
Manufactured of translucent polypropylene, these unbreakable cylinders feature blue enameled graduations from easy reading. Anti suction base eliminates problems when working on wet surfaces. Cylinders also feature unique self-draining base that allows wastewater to drain when using a labortory washing machine; helps eliminate contamination and spill re-wetting. Autoclavable.
Catalog No. NC0852346 Supplier Cole-Parmer Supplier No. 0613736
Manufactured of translucent polypropylene, these unbreakable cylinders feature blue enameled graduations from easy reading. Anti suction base eliminates problems when working on wet surfaces. Cylinders also feature unique self-draining base that allows wastewater to drain when using a labortory washing machine; helps eliminate contamination and spill re-wetting. Autoclavable.