Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Azo Biotin-Azide

Catalog No. 90089110MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-089-110MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-089-110MG Supplier Sigma Aldrich Supplier No. 90089110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Azo biotin-azide is a linker that contains a biotin moiety connected to an azide group through a spacer arm containing a diazo group that can be cleaved with 50 mM sodium dithionite solution. The biotin can be captured by streptavidin and the azide can react with terminal or strained alkynes.

Specifications

CAS 2139348-63-1
Molecular Formula C39H44ClFeNNiP2
Linear Formula C39H44ClFeNNiP2
Quantity 250 mg
Synonym SK-J002-1n
Molecular Weight (g/mol) 738.71 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.