Learn More
AdipoGen Azo-Carob Galactomannan
SDP
Supplier:  AdipoGen CDXA0221G001
Encompass_Preferred
Chemical. CAS n.a. Synthetic Remazol brilliant blue-dyed galactomannan polysaccharide. Soluble substrate for the specific measurement of endo-1,4-beta-D-mannanase/galactomannase. Used for the measurement of enzyme activity, for research, biochemical enzyme assays and in vitro diagnostic analysis.
Catalog No. 50-256-1789