Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azo Rubine, TCI America™
SDP

Catalog No. A058025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A058025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A058025G Supplier TCI America Supplier No. A058025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3567-69-9
Molecular Formula C20H12N2Na2O7S2
Molecular Weight (g/mol) 502.42
MDL Number MFCD00003978
InChI Key OJNIRNOPCQQNRY-JZFXEGLSSA-L
Synonym Brilliant Crimson Red, Carmoisine B
PubChem CID 44135716
IUPAC Name disodium 4-{2-[(2E)-1-oxo-4-sulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}naphthalene-1-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=C\C(=N/NC2=C3C=CC=CC3=C(C=C2)S([O-])(=O)=O)C(=O)C2=CC=CC=C12

Specifications

Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 502.42
Chemical Name or Material Azo Rubine
EINECSNumber (5)-1494
RTECSNumber QK1925000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.