Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azobenzene-4-sulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. A10505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10505G Supplier TCI America Supplier No. A10505G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58359-53-8
Molecular Formula C12H9ClN2O2S
Molecular Weight (g/mol) 280.73
MDL Number MFCD00024883
InChI Key UEAZPVORQYDKHZ-UHFFFAOYSA-N
Synonym 4-Phenylazobenzenesulfonyl Chloride
PubChem CID 94028
IUPAC Name 4-(2-phenyldiazen-1-yl)benzene-1-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 122°C
Quantity 5 g
UN Number 3261
Formula Weight 280.73
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Azobenzene-4-sulfonyl Chloride
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.