Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azocal A [for Calcium determination], TCI America™
SDP

Catalog No. A058225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A058225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A058225G Supplier TCI America Supplier No. A058225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1836-22-2
Molecular Formula C17H9N2Na3O9S2
Molecular Weight (g/mol) 518.35
MDL Number MFCD00046379
InChI Key SJYYQLRKEKKFSX-NETMGOMMSA-K
PubChem CID 131850130
IUPAC Name trisodium 2-{2-[(1Z)-2-oxo-3,6-disulfonato-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}benzoate
SMILES [Na+].[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1N\N=C1/C(=O)C(=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Quantity 25 g
Formula Weight 518.35
Physical Form Crystalline Powder
Chemical Name or Material Azocal A [for Calcium determination]
EINECSNumber (5)-2063&(5)-3247
RTECSNumber QJ6402600
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.