Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Azocarmine G 5 GR
SDP

Catalog No. 50980190
Encompass_Preferred

(Acid red 101, Basalan Carmine GE, and Rosinduline)
CAS #25641-18-3 C₂₈H₁₇N₃Na₂O₆S₂ F.W. 601.57
Solubility: 1.0% Water; 0.1% Alcohol; 1.75% Cellosolve; 4.5% Glycol; 0.01% Xylene

Also known as Acid red 101, Basalan Carmine GE and Rosinduline.

Catalog No. 50-980-190 Supplier Electron Microscopy Sciences Supplier No. 11150
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Acid red 101, Basalan Carmine GE, and Rosinduline)
CAS #25641-18-3 C₂₈H₁₇N₃Na₂O₆S₂ F.W. 601.57
Solubility: 1.0% Water; 0.1% Alcohol; 1.75% Cellosolve; 4.5% Glycol; 0.01% Xylene

Also known as Acid red 101, Basalan Carmine GE and Rosinduline.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.