Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azocarmine G, Certified, Thermo Scientific™

Catalog No. AC401570250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC401570250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC401570250 Supplier Thermo Scientific Chemicals Supplier No. 401570250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25641-18-3
Molecular Formula C28H18N3NaO6S2
Synonym Rosinduline
InChI Key LUERODMRBLNCFK-UHFFFAOYSA-M
SMILES C1=CC=C(C=C1)[N+]2=C3C=C(C4=C(C3=NC5=CC=CC=C52)C=CC(=C4)S(=O)(=O)[O-])NC6=CC=C(C=C6)S(=O)(=O)[O-].[Na+]
Infrared Spectrum Authentic
IUPAC Name sodium;7-phenyl-5-(4-sulfonatoanilino)benzo[a]phenazin-7-ium-3-sulfonate
Molecular Weight (g/mol) 579.577
PubChem CID 16046101
ChEBI CHEBI:87206
Lambda Max 523 to 526nm (in water)
Formula Weight 579.58
Grade Extra Pure
Color Purple or Red
Quantity 25g
Chemical Name or Material Azocarmine G
Show More Show Less
EINECSNumber 247-157-3
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.