Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azodicarboxamide, 96.0-104.0% (RT), MilliporeSigma™ Supelco™

Catalog No. 111019623 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9623 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9623 Supplier MilliporeSigma Supelco Supplier No. 7999750MG
Add to Cart
Add to Cart
Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 123-77-3
Flash Point >200°C
Linear Formula NH2 CON=NCONH2
MDL Number MFCD00007958
Melting Point 220°C to 225°C
Type Analytical Standard
Quantity 50 mg
Formula Weight 116.08
Percent Purity 96.0 to 104.0% (RT)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.