Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azomethine-H monosodium | 5941-07-1 | MFCD00042050 | 97.0% | 100 MG
SDP

Catalog No. 5000410971
Encompass_Preferred

Azomethine-H monosodium is a colour-forming reagent that is also used for boron determinations. It has been utilized for the manual determination of boron in complex borenes and boranes. Its molecular weight is 445.40 and its chemical formula is C17H12NNaO8S2. The appearance of Azomethine-H monosodium is a yellow to orange solid.

  • Reagent type: colour-forming reagent.
  • Application: used for boron determinations, including the manual determination of boron in complex borenes and boranes.
  • Physical characteristics: yellow to orange solid.
  • Solubility: soluble in DMSO (83.33 mg/mL) and H2O (5.83 mg/mL).
  • Storage: recommended storage at 4°C, sealed, away from moisture and light. Solutions in solvent should be stored at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-004-10971 Supplier Medchemexpress LLC Supplier No. HYD0797100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azomethine-H monosodium is a colour-forming reagent that is also used for boron determinations. It has been utilized for the manual determination of boron in complex borenes and boranes. Its molecular weight is 445.40 and its chemical formula is C17H12NNaO8S2. The appearance of Azomethine-H monosodium is a yellow to orange solid.

  • Reagent type: colour-forming reagent.
  • Application: used for boron determinations, including the manual determination of boron in complex borenes and boranes.
  • Physical characteristics: yellow to orange solid.
  • Solubility: soluble in DMSO (83.33 mg/mL) and H2O (5.83 mg/mL).
  • Storage: recommended storage at 4°C, sealed, away from moisture and light. Solutions in solvent should be stored at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.