Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azomethine H 97.0+%, TCI America™
SDP

Catalog No. A50165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50165G 5 g
A501625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50165G Supplier TCI America Supplier No. A50165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5941-07-1
Molecular Formula C17H14NNaO9S2
Molecular Weight (g/mol) 463.41
MDL Number MFCD00042050
InChI Key BEDIAJJRAYEWBI-TTWKNDKESA-M
Synonym 4-hydroxy-5-salicylideneamino-2,7-naphthalenedisulfonic acid sodium salt
PubChem CID 123134089
IUPAC Name 4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid;sodium
SMILES C1=CC(=CNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C(=O)C=C1.[Na].[Na]

Specifications

Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 445.39
Percent Purity ≥97.0% (T)
Chemical Name or Material Azomethine H [Spectrophotometric Reagent for B]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.