Learn More
Apexbio Technology LLC Azomycin 527-73-1 250mg
SDP
Catalog No. 5000089995
Encompass_Preferred
Azomycin (CAS 527-73-1) is a small-molecule inhibitor that interferes with electron transport pathways in aerobic Gram-positive and Gram-negative bacteria It is designed to disrupt cellular electron transfer processes thereby inhibiting bacterial respiratory function Azomycin exerts its biological activity primarily through inhibition of microbial electron transport In in vitro assays Azomycin demonstrates inhibitory activity with IC50 values typically in the micromolar concentration range though specific values may vary depending on bacterial strain and experimental conditions Based on these pharmacological properties Azomycin holds research potential in studies of bacterial metabolic pathways antimicrobial resistance mechanisms and novel antibacterial therapeutic strategies
Catalog No. 50-000-89995 Supplier Apexbio Technology LLC Supplier No. B3663250
Azomycin (CAS 527-73-1) is a small-molecule inhibitor that interferes with electron transport pathways in aerobic Gram-positive and Gram-negative bacteria It is designed to disrupt cellular electron transfer processes thereby inhibiting bacterial respiratory function Azomycin exerts its biological activity primarily through inhibition of microbial electron transport In in vitro assays Azomycin demonstrates inhibitory activity with IC50 values typically in the micromolar concentration range though specific values may vary depending on bacterial strain and experimental conditions Based on these pharmacological properties Azomycin holds research potential in studies of bacterial metabolic pathways antimicrobial resistance mechanisms and novel antibacterial therapeutic strategies