Learn More
Selleck Chemical LLC Azoramide 5mg 932986-18-0
SDP
Supplier:  Selleck Chemical LLC S83045MG
Encompass_Preferred
Azoramide is a small-molecule modulator of the unfolded protein response (UPR). It improves ER protein-folding ability and activates ER chaperone capacity to protect cells against ER stress. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8201