Learn More
Medchemexpress LLC Azoreductase/NQO1 M 10ug
SDP
Catalog No. 5000183806
Encompass_Preferred
Recombinant mouse azoreductase/NQO1 is supplied as a lyophilized, His-tagged protein expressed in Pichia pastoris, intended for biochemical characterization and enzymatic assays. The product is provided with >90% purity (reducing SDS-PAGE), an approximate molecular weight of 33 kDa, and is lyophilized from a Tris-HCl/NaCl/trehalose buffer to support stability during storage and shipment.
  • His-tagged recombinant protein produced in P. pastoris.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation in Tris-HCl, NaCl, and trehalose for stability.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O for handling.
  • Available in 5 μg, 10 μg, 50 μg, and 100 μg quantities.
  • Store at -20°C; freeze aliquots at -20°C or -80°C for long-term storage.
Catalog No. 50-001-83806 Supplier Medchemexpress LLC Supplier No. HYP7183110UG
Recombinant mouse azoreductase/NQO1 is supplied as a lyophilized, His-tagged protein expressed in Pichia pastoris, intended for biochemical characterization and enzymatic assays. The product is provided with >90% purity (reducing SDS-PAGE), an approximate molecular weight of 33 kDa, and is lyophilized from a Tris-HCl/NaCl/trehalose buffer to support stability during storage and shipment.
  • His-tagged recombinant protein produced in P. pastoris.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation in Tris-HCl, NaCl, and trehalose for stability.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O for handling.
  • Available in 5 μg, 10 μg, 50 μg, and 100 μg quantities.
  • Store at -20°C; freeze aliquots at -20°C or -80°C for long-term storage.