Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azoreductase/NQO1 M 5ug
SDP

Catalog No. 5000183342
Encompass_Preferred

Recombinant mouse Azoreductase/NQO1 enzyme expressed in Pichia pastoris with an N-terminal 6xHis tag, supplied lyophilized and formulated to preserve activity. The protein catalyzes two-electron reduction of quinones using NADH or NADPH and is suitable for biochemical assays, enzymology, and cellular redox studies.

  • Recombinant mouse enzyme expressed in Pichia pastoris.
  • N-terminal 6xHis tag for purification and detection.
  • Lyophilized formulation for extended shelf life and transport.
  • Purity greater than 90% by reducing SDS-PAGE for consistent performance.
  • Suitable for biochemical assays, kinetic studies, and redox research.
  • Stable when stored frozen; follow reconstitution and storage guidance.

Catalog No. 50-001-83342 Supplier Medchemexpress LLC Supplier No. HYP718315UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Azoreductase/NQO1 enzyme expressed in Pichia pastoris with an N-terminal 6xHis tag, supplied lyophilized and formulated to preserve activity. The protein catalyzes two-electron reduction of quinones using NADH or NADPH and is suitable for biochemical assays, enzymology, and cellular redox studies.

  • Recombinant mouse enzyme expressed in Pichia pastoris.
  • N-terminal 6xHis tag for purification and detection.
  • Lyophilized formulation for extended shelf life and transport.
  • Purity greater than 90% by reducing SDS-PAGE for consistent performance.
  • Suitable for biochemical assays, kinetic studies, and redox research.
  • Stable when stored frozen; follow reconstitution and storage guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.