Learn More
Crescent Chemical Co Inc AZORUBIN PURE
SDP
Catalog No. 50247330
Encompass_Preferred
Azorubin pure λ max. (0.001 % in water)A 1 cm/λ max. (0.001 % in water) 514 - 522min. 0.4 Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 14410.01 CAS 3567-69-9
Catalog No. 50-247-330 Supplier Crescent Chemical Co Inc Supplier No. SE14410.01
Azorubin pure λ max. (0.001 % in water)A 1 cm/λ max. (0.001 % in water) 514 - 522min. 0.4 Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 14410.01 CAS 3567-69-9