Learn More
Medchemexpress LLC HY-107321 10mg Medchemexpress, Azosemide CAS:27589-33-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10732110MG
Encompass_Preferred
Medchemexpress, HY-107321 10mg Azosemide CAS:27589-33-9 Azosemide, a sulfonamide loop diuretic, is a potent NKCC1 inhibitor with IC50s of 0.246 µM and 0.197 µM for hNKCC1A and NKCC1B, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8374