Learn More
TARGETMOL CHEMICALS INC Azosemide 200MG
SDP
Catalog No. 502880655
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Azosemide is a potent NKCC1 inhibitor (IC50s 0.246 µM and 0.197 µM for hNKCC1A and NKCC1B). Purity 98.93%
Catalog No. 50-288-0655 Supplier TARGETMOL CHEMICALS INC Supplier No. T10441200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Azosemide is a potent NKCC1 inhibitor (IC50s 0.246 µM and 0.197 µM for hNKCC1A and NKCC1B). Purity 98.93%