Learn More
ATCC Escherichia coli; JM108
Catalog No. 502381444
Encompass
Escherichia coli strain JM108 is a bacterial strain used in molecular biology research. This strain has genotype recA1, endA1, gyrA96, thi, hsdR17, supE44, relA1, Δ(lac-proAB).
Catalog No. 50-238-1444 Supplier ATCC Supplier No. 47107
Add to Cart
Add to Cart
Escherichia coli strain JM108 is a bacterial strain used in molecular biology research. This strain has genotype recA1, endA1, gyrA96, thi, hsdR17, supE44, relA1, Δ(lac-proAB).