Learn More
ATCC Bacteroides ovatus; 6 x 0.3mL
Catalog No. 502381240
Encompass
ATCC BAA-1296-MINI-PACK™ consists of 6 ready-to-use vials of ATCC BAA-1296™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-1240 Supplier ATCC Supplier No. BAA1296MINIPACK
ATCC BAA-1296-MINI-PACK™ consists of 6 ready-to-use vials of ATCC BAA-1296™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.