Learn More
Cayman Chemical AzoxystrobIn 100mg
SDP
Catalog No. 502933088
Encompass_Preferred
A broad-spectrum fungicide (LC95s = 20 = 2.9 µM); disrupts mRNA expression of antioxidant-, stress response-, and innate immune-related genes and induces the production of ROS in zebrafish larva at 0.1-100 UG/L; inhibits proliferation of KYSE-150 human esophageal squamous cell carcinoma cells (IC50 = 2.42 UG/ml after 48 hours); reduces tumor growth at 40 mg/kg per day
Catalog No. 50-293-3088 Supplier Cayman Chemical Supplier No. 24056100 MG
A broad-spectrum fungicide (LC95s = 20 = 2.9 µM); disrupts mRNA expression of antioxidant-, stress response-, and innate immune-related genes and induces the production of ROS in zebrafish larva at 0.1-100 UG/L; inhibits proliferation of KYSE-150 human esophageal squamous cell carcinoma cells (IC50 = 2.42 UG/ml after 48 hours); reduces tumor growth at 40 mg/kg per day