Learn More
Cayman Chemical Azoxystrobin, Purity: ≥98%, CAS: 131860-33-8, 50 mg
SDP
Catalog No. 502933145
Encompass_Preferred
A broad-spectrum fungicide (LC95s = 20 = 2.9 µM); disrupts mRNA expression of antioxidant-, stress response-, and innate immune-related genes and induces the production of ROS in zebrafish larva at 0.1-100 µg/L; inhibits proliferation of KYSE-150 human esophageal squamous cell carcinoma cells (IC50 = 2.42 µg/ml after 48 hours); reduces tumor growth at 40 mg/kg per day
Catalog No. 50-293-3145 Supplier Cayman Chemical Supplier No. 2405650 MG
A broad-spectrum fungicide (LC95s = 20 = 2.9 µM); disrupts mRNA expression of antioxidant-, stress response-, and innate immune-related genes and induces the production of ROS in zebrafish larva at 0.1-100 µg/L; inhibits proliferation of KYSE-150 human esophageal squamous cell carcinoma cells (IC50 = 2.42 µg/ml after 48 hours); reduces tumor growth at 40 mg/kg per day