Learn More
Medchemexpress LLC AZP-531 | 1088543-62-7 | 99.8% | 962.02 | 1 MG
SDP
Catalog No. 5000425815
Encompass_Preferred
AZP-531 is an analogue of unacylated ghrelin for research use, designed to improve glycaemic control and reduce weight. It shows survival effects on pancreatic β-cells and human pancreatic islets, and is highly stable in human plasma in vitro with no degradation after one day at 37°C.
  • Exhibits survival effects on pancreatic β-cells and human pancreatic islets.
  • Stable in human plasma in vitro at 37°C for one day.
  • Prevents high-fat diet-induced weight gain, proinflammatory effects, and prediabetic metabolic state in mice.
  • Stimulates mitochondrial function markers in brown adipose tissue.
  • Well-tolerated in clinical trials; improves glucose without increasing insulin.
  • Reduces mean body weight and HbA1c in clinical trials.
Catalog No. 50-004-25815 Supplier Medchemexpress LLC Supplier No. HYP02311MG
AZP-531 is an analogue of unacylated ghrelin for research use, designed to improve glycaemic control and reduce weight. It shows survival effects on pancreatic β-cells and human pancreatic islets, and is highly stable in human plasma in vitro with no degradation after one day at 37°C.
  • Exhibits survival effects on pancreatic β-cells and human pancreatic islets.
  • Stable in human plasma in vitro at 37°C for one day.
  • Prevents high-fat diet-induced weight gain, proinflammatory effects, and prediabetic metabolic state in mice.
  • Stimulates mitochondrial function markers in brown adipose tissue.
  • Well-tolerated in clinical trials; improves glucose without increasing insulin.
  • Reduces mean body weight and HbA1c in clinical trials.