Learn More
Medchemexpress LLC AZP-531 | 1088543-62-7 | 99.8% | 962.02 | 10 MG
SDP
Catalog No. 5000425897
Encompass_Preferred
AZP-531 is an analogue of unacylated ghrelin designed to improve glycaemic control and reduce weight. It exerts survival effects on pancreatic b-cells and human pancreatic islets comparable to unacylated ghrelin (UAG). It is very stable in human plasma in vitro with no degradation observed after 1 day of incubation at 37°C.
  • Prevents body weight increase caused by a high-fat diet in mice
  • Prevents high-fat diet-induced proinflammatory effects
  • Stimulates mitochondrial function markers in brown adipose tissue
  • Prevents the development of a prediabetic metabolic state
  • Prevents a high-fat diet-induced increase in acyl ghrelin levels
  • Significantly improves glucose concentrations without increasing insulin levels
  • Decreases mean body weight by 2.6 kg (vs 0.8 kg for placebo)
  • Reduces HbA1c by 0.4% (vs 0.2% for placebo)
Catalog No. 50-004-25897 Supplier Medchemexpress LLC Supplier No. HYP023110MG
AZP-531 is an analogue of unacylated ghrelin designed to improve glycaemic control and reduce weight. It exerts survival effects on pancreatic b-cells and human pancreatic islets comparable to unacylated ghrelin (UAG). It is very stable in human plasma in vitro with no degradation observed after 1 day of incubation at 37°C.
  • Prevents body weight increase caused by a high-fat diet in mice
  • Prevents high-fat diet-induced proinflammatory effects
  • Stimulates mitochondrial function markers in brown adipose tissue
  • Prevents the development of a prediabetic metabolic state
  • Prevents a high-fat diet-induced increase in acyl ghrelin levels
  • Significantly improves glucose concentrations without increasing insulin levels
  • Decreases mean body weight by 2.6 kg (vs 0.8 kg for placebo)
  • Reduces HbA1c by 0.4% (vs 0.2% for placebo)