Learn More
Medchemexpress LLC AZT triphosphate tet 10mg | 106060-92-8 | 10 MG
SDP
Catalog No. 5000227342
Encompass_Preferred
AZT triphosphate tetraammonium is the tetraammonium salt of the active triphosphate metabolite of zidovudine (AZT). It is used as a biochemical reagent in virology and nucleotide metabolism studies where the active nucleotide analog is required. The product is supplied with a certificate of analysis and safety data sheet and is intended for research use only.
  • Active triphosphate metabolite of zidovudine (AZT) for biochemical assays.
  • High chemical purity suitable for analytical and cellular studies.
  • Available as a solid in multiple milligram sizes and as a 10 mM solution in DMSO.
  • Supplied with certificate of analysis and safety data sheet for quality and handling information.
  • Recommended storage: sealed at -20°C; in solvent store at -80°C for up to 6 months.
Catalog No. 50-002-27342 Supplier Medchemexpress LLC Supplier No. HY116364B10MG
AZT triphosphate tetraammonium is the tetraammonium salt of the active triphosphate metabolite of zidovudine (AZT). It is used as a biochemical reagent in virology and nucleotide metabolism studies where the active nucleotide analog is required. The product is supplied with a certificate of analysis and safety data sheet and is intended for research use only.
  • Active triphosphate metabolite of zidovudine (AZT) for biochemical assays.
  • High chemical purity suitable for analytical and cellular studies.
  • Available as a solid in multiple milligram sizes and as a 10 mM solution in DMSO.
  • Supplied with certificate of analysis and safety data sheet for quality and handling information.
  • Recommended storage: sealed at -20°C; in solvent store at -80°C for up to 6 months.