Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AZT triphosphate tet 10mM 1mL | 106060-92-8 | 579.37 | C10H16N5O13P3•4NH4 | 1 ML
SDP

Catalog No. 5000229467
Encompass_Preferred

AZT triphosphate (tetraammonium) is the 5′-triphosphate metabolite of zidovudine (AZT), supplied as a 10 mM solution in a 1 mL vial for biochemical and virology research such as reverse transcriptase inhibition and nucleotide incorporation assays.

  • Supplied as the tetraammonium salt
  • Concentration 10 mM in 1 mL vial
  • Purity 99.25%
  • Molecular formula C10H16N5O13P3 • 4NH4
  • Molecular weight 579.37 (tetraammonium salt)
  • Intended for biochemical assays and reverse transcriptase studies

Catalog No. 50-002-29467 Supplier Medchemexpress LLC Supplier No. HY116364B10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AZT triphosphate (tetraammonium) is the 5′-triphosphate metabolite of zidovudine (AZT), supplied as a 10 mM solution in a 1 mL vial for biochemical and virology research such as reverse transcriptase inhibition and nucleotide incorporation assays.

  • Supplied as the tetraammonium salt
  • Concentration 10 mM in 1 mL vial
  • Purity 99.25%
  • Molecular formula C10H16N5O13P3 • 4NH4
  • Molecular weight 579.37 (tetraammonium salt)
  • Intended for biochemical assays and reverse transcriptase studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.