Learn More
Medchemexpress LLC AZT triphosphate tet 1mg | 106060-92-8 | 1 MG
SDP
Catalog No. 5000229851
Encompass_Preferred
AZT triphosphate tetraammonium is the tetraammonium salt of the active triphosphate metabolite of zidovudine (AZT). Supplied as an off-white to light yellow solid, it is used as a biochemical reagent and reference standard in studies of viral polymerase activity and nucleotide metabolism.
  • Active triphosphate metabolite of zidovudine with antiretroviral activity.
  • Supplied as a stable solid suitable for biochemical assays and standards.
  • High purity (99.25%) to support analytical and research applications.
  • Store sealed at -20°C; in solution store at -80°C for long-term stability.
  • Available in small quantities (for example, 1 mg) and larger sizes for research use.
Catalog No. 50-002-29851 Supplier Medchemexpress LLC Supplier No. HY116364B1MG
AZT triphosphate tetraammonium is the tetraammonium salt of the active triphosphate metabolite of zidovudine (AZT). Supplied as an off-white to light yellow solid, it is used as a biochemical reagent and reference standard in studies of viral polymerase activity and nucleotide metabolism.
  • Active triphosphate metabolite of zidovudine with antiretroviral activity.
  • Supplied as a stable solid suitable for biochemical assays and standards.
  • High purity (99.25%) to support analytical and research applications.
  • Store sealed at -20°C; in solution store at -80°C for long-term stability.
  • Available in small quantities (for example, 1 mg) and larger sizes for research use.