Learn More
Alkali Scientific Aztreonam, 1 G
SDP
Catalog No. 5000660445
Encompass_Preferred
Aztreonam (1 g) is a monobactam antibiotic used in laboratory-grade antibacterial research. It is highly effective against Gram-negative bacteria and operates by inhibiting cell wall synthesis. This antibiotic is widely utilized in research to study the efficacy of antibacterial agents, particularly in drug resistance and antibacterial therapy studies. Aztreonam's mechanism of action makes it a crucial reagent in research on antibiotic resistance, pharmacokinetics, and pharmacodynamics. It is also commonly used in microbiological testing to analyze bacterial responses and resistance patterns in clinical and laboratory settings.
Catalog No. 50-006-60445 Supplier Alkali Scientific Supplier No. AZT003CP
Aztreonam (1 g) is a monobactam antibiotic used in laboratory-grade antibacterial research. It is highly effective against Gram-negative bacteria and operates by inhibiting cell wall synthesis. This antibiotic is widely utilized in research to study the efficacy of antibacterial agents, particularly in drug resistance and antibacterial therapy studies. Aztreonam's mechanism of action makes it a crucial reagent in research on antibiotic resistance, pharmacokinetics, and pharmacodynamics. It is also commonly used in microbiological testing to analyze bacterial responses and resistance patterns in clinical and laboratory settings.