Learn More
Research Products International Corp Aztreonam, 1 G
SDP
Supplier:  Research Products International Corp A721001.0
Encompass_Preferred
Aztreonam is a synthetic monocyclic beta-lactam antibiotic, with the nucleus based on a simpler monobactam isolated from Chromobacterium violaceum.
Catalog No. 50-197-7086