Learn More
TARGETMOL CHEMICALS INC Aztreonam 200MG
SDP
Catalog No. 502880613
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Aztreonam (SQ-26,776), a monocyclic beta-lactam antibiotic originally isolated from Chromobacterium violaceum, possesses bactericidal activity. Purity 99.42%
Catalog No. 50-288-0613 Supplier TARGETMOL CHEMICALS INC Supplier No. T1029200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Aztreonam (SQ-26,776), a monocyclic beta-lactam antibiotic originally isolated from Chromobacterium violaceum, possesses bactericidal activity. Purity 99.42%