Learn More
Alkali Scientific Aztreonam, 25 Mg
SDP
Catalog No. 5000660861
Encompass_Preferred
Aztreonam is a monobactam antibiotic used in antibacterial research. This 25mg laboratory-grade sample is used for studying bacterial resistance mechanisms and testing the efficacy of monobactam antibiotics in inhibiting bacterial growth. Aztreonam is particularly effective against Gram-negative bacteria, making it an important compound for research on infectious diseases caused by these organisms. It is also utilized in laboratory studies to explore the antibacterial properties of monobactams in combination therapies. This 25mg quantity is sufficient for several antimicrobial studies and assay testing in microbiological research.
Catalog No. 50-006-60861 Supplier Alkali Scientific Supplier No. AZT001CP
Aztreonam is a monobactam antibiotic used in antibacterial research. This 25mg laboratory-grade sample is used for studying bacterial resistance mechanisms and testing the efficacy of monobactam antibiotics in inhibiting bacterial growth. Aztreonam is particularly effective against Gram-negative bacteria, making it an important compound for research on infectious diseases caused by these organisms. It is also utilized in laboratory studies to explore the antibacterial properties of monobactams in combination therapies. This 25mg quantity is sufficient for several antimicrobial studies and assay testing in microbiological research.