Learn More
Medchemexpress LLC Aztreonam | 78110-38-0 | 99.6% | 200 MG
SDP
Catalog No. 5000450938
Encompass_Preferred
Aztreonam (SQ-26,776) is a synthetic monocyclic beta-lactam antibiotic that exhibits a very high affinity for penicillin-binding protein 3 (PBP-3). It functions by inhibiting mucopeptide synthesis within the bacterial cell wall, thereby preventing peptidoglycan crosslinking. While resistant to some beta-lactamases, it is inactivated by extended-spectrum beta-lactamases. Aztreonam demonstrates no useful activity against gram-positive or anaerobic microorganisms.
  • Inhibits mucopeptide synthesis in bacterial cell walls
  • Blocks peptidoglycan crosslinking
  • High affinity for penicillin-binding protein 3 (PBP-3)
  • Resistant to certain beta-lactamases
  • Bactericidal properties
Catalog No. 50-004-50938 Supplier Medchemexpress LLC Supplier No. HYB0129200MG
Aztreonam (SQ-26,776) is a synthetic monocyclic beta-lactam antibiotic that exhibits a very high affinity for penicillin-binding protein 3 (PBP-3). It functions by inhibiting mucopeptide synthesis within the bacterial cell wall, thereby preventing peptidoglycan crosslinking. While resistant to some beta-lactamases, it is inactivated by extended-spectrum beta-lactamases. Aztreonam demonstrates no useful activity against gram-positive or anaerobic microorganisms.
  • Inhibits mucopeptide synthesis in bacterial cell walls
  • Blocks peptidoglycan crosslinking
  • High affinity for penicillin-binding protein 3 (PBP-3)
  • Resistant to certain beta-lactamases
  • Bactericidal properties