Learn More
Medchemexpress LLC Aztreonam-d6 | 1127452-94-1 | 99.3% | 1 MG
SDP
Catalog No. 5000450466
Encompass_Preferred
Aztreonam-d6 is deuterium labeled Aztreonam. Aztreonam (SQ-26,776) is a synthetic monocyclic beta-lactam antibiotic that has a very high affinity for penicillin-binding protein 3 (PBP-3). It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled aztreonam
  • Synthetic monocyclic beta-lactam antibiotic
  • High affinity for penicillin-binding protein 3 (PBP-3)
  • Useful as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
Catalog No. 50-004-50466 Supplier Medchemexpress LLC Supplier No. HYB0129S1MG
Aztreonam-d6 is deuterium labeled Aztreonam. Aztreonam (SQ-26,776) is a synthetic monocyclic beta-lactam antibiotic that has a very high affinity for penicillin-binding protein 3 (PBP-3). It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled aztreonam
  • Synthetic monocyclic beta-lactam antibiotic
  • High affinity for penicillin-binding protein 3 (PBP-3)
  • Useful as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only