Learn More
Medchemexpress LLC Aztreonam E-isomer | 99341-02-3 | MFCD09028013 | 98.7% | C13H17N5O8S2 | 10MG
SDP
Catalog No. 5000258801
Encompass_Preferred
(E)-Aztreonam is the E-isomer of the monocyclic β-lactam antibiotic aztreonam, provided as a research-grade powder for in vitro studies of gram-negative bacterial infections and related assays. The material is characterized by a molecular weight of 435.43 and a reported purity of about 98.7%; follow recommended storage to preserve stability.
  • Monocyclic β-lactam antibiotic, E-isomer
  • Reported purity approximately 98.7%
  • Intended for in vitro research on gram-negative pathogens
  • Supplied as a powder with defined storage conditions
  • Molecular formula C13H17N5O8S2; molecular weight 435.43
  • Available in small milligram quantities suitable for analytical use
Catalog No. 50-002-58801 Supplier Medchemexpress LLC Supplier No. HY13650210MG
(E)-Aztreonam is the E-isomer of the monocyclic β-lactam antibiotic aztreonam, provided as a research-grade powder for in vitro studies of gram-negative bacterial infections and related assays. The material is characterized by a molecular weight of 435.43 and a reported purity of about 98.7%; follow recommended storage to preserve stability.
  • Monocyclic β-lactam antibiotic, E-isomer
  • Reported purity approximately 98.7%
  • Intended for in vitro research on gram-negative pathogens
  • Supplied as a powder with defined storage conditions
  • Molecular formula C13H17N5O8S2; molecular weight 435.43
  • Available in small milligram quantities suitable for analytical use